Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 3-(pyridin-2-ylamino)propanoate 95%

Supplier: AMBEED, INC
Warning

A415414-1G A415414-250MG A415414-5G
76725-206EA 53 USD InStock
76725-206 76725-208 76725-210
Methyl 3-(pyridin-2-ylamino)propanoate 95%
Methyl 3-(pyridin-2-ylamino)propanoate
Precaución: for Research Use Only


Formula: C₉H₁₂N₂O₂
MDL Number: MFCD02708568
CAS Number: 55364-85-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty