Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Iodo-4-methoxypyrimidine 95%

Supplier: AMBEED, INC
Warning

A420246-1G A420246-5G
76725-228EA 24 USD InStock
76725-228 76725-230
2-Iodo-4-methoxypyrimidine 95%
2-Iodo-4-methoxypyrimidine
Precaución: for Research Use Only


Formula: C₅H₅IN₂O
MDL Number: MFCD08275719
CAS Number: 262353-35-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty