Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium-3-fluoro-4-methoxyphenyltrifluoroborate 95%

Supplier: AMBEED, INC
Warning

A424402-1G A424402-250MG A424402-5G
76725-260EA 19 USD InStock
76725-260 76725-262 76725-264
Potassium-3-fluoro-4-methoxyphenyltrifluoroborate 95%
Potassium 3-fluoro-4-methoxyphenyltrifluoroborate
Precaución: for Research Use Only


Formula: C₇H₆BF₄KO
MDL Number: MFCD04115757
CAS Number: 850623-62-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty