Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzo[1,3]dioxole-5-carboxamidoxime 95%

Supplier: AMBEED, INC
Warning

A428547-1G A428547-250MG A428547-5G
76725-302EA 23 USD InStock
76725-302 76725-304 76725-306
Benzo[1,3]dioxole-5-carboxamidoxime 95%
Benzo[1,3]dioxole-5-carboxamidoxime
Precaución: for Research Use Only


Formula: C₈H₈N₂O₃
MDL Number: MFCD00119015
CAS Number: 4720-72-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty