Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hexyltrimethoxysilane 98%

Supplier: AMBEED, INC
Warning

A428690-100G A428690-25G A428690-5G
76725-324EA 52 USD InStock
76725-324 76725-326 76725-328
Hexyltrimethoxysilane 98%
Hexyltrimethoxysilane
Precaución: for Research Use Only


Formula: C₉H₂₂O₃Si
MW: 206.36 g/mol
MDL Number: MFCD00039801
CAS Number: 3069-19-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty