Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Benzyloxycarbonyl-4-oxo-L-proline 98%

Supplier: AMBEED, INC
Warning

A429071-1G A429071-25G A429071-5G
76725-330EA 44 USD InStock
76725-330 76725-332 76725-334
1-Benzyloxycarbonyl-4-oxo-L-proline 98%
1-Benzyloxycarbonyl-4-oxo-L-proline
Precaución: for Research Use Only


Formula: C₁₃H₁₃NO₅
MW: 263.249 g/mol
MDL Number: MFCD07367987
CAS Number: 64187-47-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty