Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N-Bis(2-hydroxyethyl)-3-methoxyaniline 98%

Supplier: AMBEED, INC
Danger

A440208-1G A440208-5G
76725-452EA 24 USD InStock
76725-452 76725-454
N,N-Bis(2-hydroxyethyl)-3-methoxyaniline 98%
N,N-Bis(2-hydroxyethyl)-3-methoxyaniline
Precaución: for Research Use Only


Formula: C₁₁H₁₇NO₃
MDL Number: MFCD02683045
CAS Number: 17126-75-9
UN: 3259
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty