Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-5-amino-2-methoxybenzoate 97%

Supplier: AMBEED, INC
Warning

A442145-1G A442145-250MG A442145-25G A442145-5G
76725-464EA 67 USD InStock
76725-464 76725-466 76725-468 76725-470
Methyl-5-amino-2-methoxybenzoate 97%
Methyl-5-amino-2-methoxybenzoate
Precaución: for Research Use Only


Formula: C₉H₁₁NO₃
MW: 181.19 g/mol
MDL Number: MFCD08458835
CAS Number: 22802-67-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty