Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nilotinib 98%

Supplier: AMBEED, INC
Warning

A448494-100MG A448494-1MG A448494-5G A448494-250MG
76725-548EA 23 USD InStock
76725-548 76725-550 76725-552 77669-014
Nilotinib 98%
Nilotinib
Precaución: for Research Use Only


Formula: C₂₈H₂₂F₃N₇O
MW: 529.5 g/mol
MDL Number: MFCD09833716
CAS Number: 641571-10-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty