Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Carbamoylphenylboronic acid pinacol ester 95%

Supplier: AMBEED, INC
Warning

A452926-250MG A452926-25G A452926-5G
76725-584EA 9 USD InStock
76725-584 76725-586 76725-588
4-Carbamoylphenylboronic acid pinacol ester 95%
4-Carbamoylphenylboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₃H₁₈BNO₃
MDL Number: MFCD05663887
CAS Number: 179117-44-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty