Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium-2-hydroxyphenyltrifluoroborate 95%

Supplier: AMBEED, INC
Warning

A458848-1G A458848-25G A458848-5G
76725-670EA 14 USD InStock
76725-670 76725-672 76725-674
Potassium-2-hydroxyphenyltrifluoroborate 95%
Potassium 2-hydroxyphenyltrifluoroborate
Precaución: for Research Use Only


Formula: C₆H₅BF₃KO
MDL Number: MFCD07781243
CAS Number: 850313-92-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty