Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Ethynyl-4-phenoxybenzene 97%

Supplier: AMBEED, INC
Danger

A459036-1G A459036-250MG A459036-5G
76725-682EA 76 USD InStock
76725-682 76725-684 76725-686
1-Ethynyl-4-phenoxybenzene 97%
1-Ethynyl-4-phenoxybenzene
Precaución: for Research Use Only


Formula: C₁₄H₁₀O
MDL Number: MFCD03839992
CAS Number: 4200-06-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty