Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium nitroprusside dihydrate 99%

Supplier: AMBEED, INC
Danger

A464682-250MG A464682-25G A464682-50MG A464682-5G
76725-728EA 6 USD InStock
76725-728 76725-730 76725-732 76725-734
Sodium nitroprusside dihydrate 99%
Sodium nitroprusside dihydrate
Precaución: for Research Use Only


Formula: Na₂[Fe(CN)₅NO]·2H₂O
MW: 297.95 g/mol
MDL Number: MFCD00149192
CAS Number: 13755-38-9
UN: 3288
ADR: 6.1 Ⅲ
Merck Index: 13,08722
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty