Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-Ethyl-2-acetamido-3-phenylpropanoate 98%

Supplier: AMBEED, INC
Warning

A466507-1G A466507-250MG A466507-25G A466507-5G
76725-758EA 11 USD InStock
76725-758 76725-760 76725-762 76725-764
(S)-Ethyl-2-acetamido-3-phenylpropanoate 98%
(S)-Ethyl-2-acetamido-3-phenylpropanoate
Precaución: for Research Use Only


Formula: C₁₃H₁₇NO₃
MW: 235.283 g/mol
MDL Number: MFCD00026885
CAS Number: 2361-96-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty