Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-Ethyl 2-acetamido-3-mercaptopropanoate 95%

Supplier: AMBEED, INC
Danger

A467353-250MG A467353-1G A467353-100MG
76725-766EA 33 USD InStock
76725-766 77572-472 77669-104
(R)-Ethyl 2-acetamido-3-mercaptopropanoate 95%
(R)-Ethyl 2-acetamido-3-mercaptopropanoate
Precaución: for Research Use Only


Formula: C₇H₁₃NO₃S
MDL Number: MFCD09954466
CAS Number: 59587-09-6
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty