Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Nitrophenylboronic acid pinacol ester 98%

Supplier: AMBEED, INC
Warning

A469791-1G A469791-25G A469791-5G
76725-786EA 10 USD InStock
76725-786 76725-788 76725-790
3-Nitrophenylboronic acid pinacol ester 98%
3-Nitrophenylboronic acid pinacol ester


Formula: C₁₂H₁₆BNO₄
MDL Number: MFCD05663879
CAS Number: 68716-48-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty