Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dodecyltriethoxysilane 98%

Supplier: AMBEED, INC
Warning

A474314-100G A474314-25G A474314-500G
76725-814EA 10 USD InStock
76725-814 76725-816 76725-818
Dodecyltriethoxysilane 98%
Dodecyltriethoxysilane
Precaución: for Research Use Only


Formula: C₁₈H₄₀O₃Si
MDL Number: MFCD00048437
CAS Number: 18536-91-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty