Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N-Dimethylformamide di-tert-butyl acetal 90%

Supplier: AMBEED, INC
Danger

A482910-1G A482910-250MG A482910-5G A482910-25G
76725-910EA 8 USD InStock
76725-910 76725-912 76725-914 77594-504
N,N-Dimethylformamide di-tert-butyl acetal 90%
N,N-Dimethylformamide di-tert-butyl acetal
Precaución: for Research Use Only


Formula: C₁₁H₂₅NO₂
MW: 203.32 g/mol
MDL Number: MFCD00015002
CAS Number: 36805-97-7
UN: 1993
ADR: 3 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty