Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(2-Pyridyl)benzaldehyde 96%

Supplier: AMBEED, INC
Warning

A485205-1G A485205-250MG A485205-5G
76725-928EA 39 USD InStock
76725-928 76725-930 76725-932
3-(2-Pyridyl)benzaldehyde 96%
3-(2-Pyridyl)benzaldehyde
Precaución: for Research Use Only


Formula: C₁₂H₉NO
MDL Number: MFCD02684103
CAS Number: 85553-53-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty