Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N,N'-Trimethylethylenediamine 98%

Supplier: AMBEED, INC
Danger

A486275-10G A486275-1G A486275-25G A486275-5G A486275-100G
76725-934EA 15 USD InStock
76725-934 76725-936 76725-938 76725-940 77669-172
N,N,N'-Trimethylethylenediamine 98%
N,N,N'-Trimethylethylenediamine
Precaución: for Research Use Only


Formula: C₅H₁₄N₂
MW: 102.18 g/mol
Boiling Pt: 116
MDL Number: MFCD00014874
CAS Number: 142-25-6
UN: 2733
ADR: 3,8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty