Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Oxalylglycine 98%

Supplier: AMBEED, INC
Danger

A487377-100MG A487377-250MG A487377-1G A487377-5G
76725-948EA 40 USD InStock
76725-948 76725-950 77602-106 77669-176
N-Oxalylglycine 98%
N-Oxalylglycine
Precaución: for Research Use Only


Formula: C₄H₅NO₅
MDL Number: MFCD00913253
CAS Number: 5262-39-5
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty