Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(2-Bromobenzyl)-2-chloro-N-ethylethanamine hydrochloride 95%

Supplier: AMBEED, INC
Danger

A494835-10MG A494835-1MG A494835-5MG
76725-978EA 75 USD InStock
76725-978 76725-980 76725-982
N-(2-Bromobenzyl)-2-chloro-N-ethylethanamine hydrochloride 95%
N-(2-Bromobenzyl)-2-chloro-N-ethylethanamine hydrochloride
Precaución: for Research Use Only


Formula: C₁₁H₁₆BrCl₂N
MDL Number: MFCD00043241
CAS Number: 40616-75-9
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty