Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2-Chlorophenyl)urea 95%

Supplier: AMBEED, INC
Warning

A495137-100MG A495137-1G A495137-25G
76725-984EA 7 USD InStock
76725-984 76725-986 76725-988
1-(2-Chlorophenyl)urea 95%
1-(2-Chlorophenyl)urea
Precaución: for Research Use Only


Formula: C₇H₇ClN₂O
MDL Number: MFCD00041316
CAS Number: 114-38-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty