Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxy-3-methoxyphenylboronic acid pinacol ester 98%

Supplier: AMBEED, INC
Warning

A503594-1G A503594-250MG A503594-5G
76726-048EA 41 USD InStock
76726-048 76726-050 76726-052
4-Hydroxy-3-methoxyphenylboronic acid pinacol ester 98%
4-Hydroxy-3-methoxyphenylboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₃H₁₉BO₄
MDL Number: MFCD02093723
CAS Number: 269410-22-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty