Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(4-Chlorophenyl)acrylamide 95% stabilized

Supplier: AMBEED, INC
Warning

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A504407-100MG A504407-1G A504407-250MG A504407-5G
76726-066EA 45 USD InStock
76726-066 76726-068 76726-070 76726-072
N-(4-Chlorophenyl)acrylamide 95% stabilized
N-(4-Chlorophenyl)acrylamide
Precaución: for Research Use Only


Formula: C₉H₈ClNO
MDL Number: MFCD00045201
CAS Number: 5453-48-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty