Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Disodium pamidronate pentahydrate 98%

Supplier: AMBEED, INC
Warning

A505899-1G A505899-25G A505899-5G
76726-074EA 32 USD InStock
76726-074 76726-076 76726-078
Disodium pamidronate pentahydrate 98%
Disodium pamidronate pentahydrate
Precaución: for Research Use Only


Formula: C₃H₁₉NNa₂O₁₂P₂
MW: 369.11 g/mol
MDL Number: MFCD01938386
CAS Number: 109552-15-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty