Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

O-tert-Butylhydroxylamine hydrochloride 98%

Supplier: AMBEED, INC
Danger

A507347-1G A507347-250MG A507347-25G A507347-5G
76726-086EA 12 USD InStock
76726-086 76726-088 76726-090 76726-092
O-tert-Butylhydroxylamine hydrochloride 98%
O-tert-Butylhydroxylamine hydrochloride


Formula: C₄H₁₂ClNO
MDL Number: MFCD00043272
CAS Number: 39684-28-1
UN: 1325
ADR: 4.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty