Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Phenoxathiin-4-boronic acid 95%

Supplier: AMBEED, INC
Warning

A516427-1G A516427-250MG A516427-5G
76726-170EA 55 USD InStock
76726-170 76726-172 77669-294
Phenoxathiin-4-boronic acid 95%
Phenoxathiin-4-boronic acid
Precaución: for Research Use Only


Formula: C₁₂H₉BO₃S
MW: 244.08 g/mol
MDL Number: MFCD01605731
CAS Number: 100124-07-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty