Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,4-Bis(dimethylamino)butane 98%

Supplier: AMBEED, INC
Danger

A523647-100MG A523647-250MG A523647-5G
76726-220EA 6 USD InStock
76726-220 76726-222 77669-318
1,4-Bis(dimethylamino)butane 98%
1,4-Bis(dimethylamino)butane
Precaución: for Research Use Only


Formula: C₈H₂₀N₂
MW: 144.26 g/mol
Boiling Pt: 166
MDL Number: MFCD00008338
CAS Number: 111-51-3
UN: 2734
ADR: 8,3 Ⅱ
Merck Index: 12,09365
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty