Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl pent-2-enoate 95%

Supplier: AMBEED, INC
Danger

A525889-10G A525889-1G
76726-236EA 55 USD InStock
76726-236 77669-330
Methyl pent-2-enoate 95%
Methyl pent-2-enoate
Precaución: for Research Use Only


Formula: C₆H₁₀O₂
MW: 114.142 g/mol
MDL Number: MFCD00137611
CAS Number: 818-59-7
UN: 1993
ADR: 3 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty