Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Methylcycloheptanamine hydrochloride 97%

Supplier: AMBEED, INC
Warning

A527584-100MG A527584-250MG A527584-5G
76726-246EA 13 USD InStock
76726-246 76726-248 76726-250
N-Methylcycloheptanamine hydrochloride 97%
N-Methylcycloheptanamine hydrochloride
Precaución: for Research Use Only


Formula: C₈H₁₈ClN
MDL Number: MFCD13186330
CAS Number: 1255718-16-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty