Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N-Diethyl-2-phenylacetamide 97%

Supplier: AMBEED, INC
Warning

A539972-100G A539972-10G A539972-25G A539972-5G
76726-354EA 1334 USD InStock
76726-354 76726-356 76726-358 76726-360
N,N-Diethyl-2-phenylacetamide 97%
N,N-Diethyl-2-phenylacetamide
Precaución: for Research Use Only


Formula: C₁₂H₁₇NO
MW: 191.26 g/mol
MDL Number: MFCD00192031
CAS Number: 2431-96-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty