Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-5-chlorovalerate 95%

Supplier: AMBEED, INC
Warning

A544063-100G A544063-10G A544063-25G A544063-500G
76726-398EA 63 USD InStock
76726-398 76726-400 76726-402 76726-404
Methyl-5-chlorovalerate 95%
Methyl 5-chlorovalerate
Precaución: for Research Use Only


Formula: C₆H₁₁ClO₂
MDL Number: MFCD00065274
CAS Number: 14273-86-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty