Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Perfluoroheptanoic acid 98%

Supplier: AMBEED, INC
Danger

A545426-100MG A545426-250MG A545426-1G
76726-418EA 6 USD InStock
76726-418 76726-420 77669-410
Perfluoroheptanoic acid 98%
Perfluoroheptanoic acid
Precaución: for Research Use Only


Formula: C₇HF₁₃O₂
MW: 364.06
MDL Number: MFCD00039604
CAS Number: 375-85-9
UN: 3265
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty