Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pelargonic acid 95%

Supplier: AMBEED, INC
Warning

A551924-25G A551924-500G
76726-464EA 9 USD InStock
76726-464 76726-466
Pelargonic acid 95%
Pelargonic acid
Precaución: for Research Use Only


Formula: C₉H₁₈O₂
MW: 158.24 g/mol
Boiling Pt: 253
Melting Pt: 12
Flash Pt: 133
MDL Number: MFCD00004433
CAS Number: 112-05-0
Merck Index: 12,07198
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty