Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium stearate 98%

Supplier: AMBEED, INC
Danger

A557433-1000G A557433-500G A557433-100G
76726-492EA 5.31 USD InStock
76726-492 77602-366 77602-368 77669-474
Sodium stearate 98%
Sodium stearate
Precaución: for Research Use Only


Formula: C₁₈H₃₅NaO₂
MW: 306.46 g/mol
Melting Pt: 288
MDL Number: MFCD00036404
CAS Number: 822-16-2
UN: 3077
ADR: 9 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty