Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Palmitoleic acid 98+%

Supplier: AMBEED, INC
Warning

A559345-1G A559345-5G A559345-250MG A559345-25G
76726-518EA 30 USD InStock
76726-518 77569-078 77602-380 77669-480
Palmitoleic acid 98+%
Palmitoleic acid
Precaución: For research use only.


Formula: C₁₆H₃₀O₂
MW: 254.41 g/mol
MDL Number: MFCD00004437
CAS Number: 373-49-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty