Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-fluoro-4-methoxybenzoate 98%

Supplier: AMBEED, INC
Warning

A559532-1G A559532-25G A559532-5G
76726-520EA 14 USD InStock
76726-520 76726-522 76726-524
Methyl-3-fluoro-4-methoxybenzoate 98%
Methyl-3-fluoro-4-methoxybenzoate
Precaución: for Research Use Only


Formula: C₉H₉FO₃
MDL Number: MFCD00215839
CAS Number: 369-30-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty