Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Pyridineboronic acid hydrochloride 98%

Supplier: AMBEED, INC
Warning

A561077-100G A561077-25G A561077-5G
76726-538EA 308 USD InStock
76726-538 76726-540 76726-542
4-Pyridineboronic acid hydrochloride 98%
4-Pyridineboronic acid hydrochloride
Precaución: for Research Use Only


Formula: C₅H₇BClNO₂
MDL Number: MFCD06201888
CAS Number: 913835-65-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty