Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-mercaptopropionate 95%

Supplier: AMBEED, INC
Danger

A564637-100G A564637-25G A564637-500G
76726-558EA 63 USD InStock
76726-558 76726-560 76726-562
Methyl-3-mercaptopropionate 95%
Methyl-3-mercaptopropionate
Precaución: for Research Use Only


Formula: C₄H₈O₂S
MW: 120.17
Boiling Pt: 54
Melting Pt: -60
Density: 1.085
MDL Number: MFCD00004895
CAS Number: 2935-90-2
UN: 2810
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty