Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Methoxy-N-methyltrifluoroacetamide 97%

Supplier: AMBEED, INC
Danger

A569817-10G A569817-1G A569817-25G A569817-5G
76726-630EA 15 USD InStock
76726-630 76726-632 76726-634 76726-636
N-Methoxy-N-methyltrifluoroacetamide 97%
N-Methoxy-N-methyltrifluoroacetamide
Precaución: for Research Use Only


Formula: C₄H₆F₃NO₂
MW: 157.09 g/mol
MDL Number: MFCD00077565
CAS Number: 104863-67-4
UN: 1992
ADR: 3,6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty