Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nickel(II) acetylacetonate dihydrate 98%

Supplier: AMBEED, INC
Warning

A578199-100G A578199-25G A578199-5G
76726-718EA 44 USD InStock
76726-718 76726-720 76726-722
Nickel(II) acetylacetonate dihydrate 98%
Nickel(II) acetylacetonate dihydrate
Precaución: for Research Use Only


Formula: C₁₀H₁₈NiO₆
MW: 292.94 g/mol
MDL Number: MFCD00149059
CAS Number: 14363-16-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty