Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Furfuryl methyl disulfide 98%

Supplier: AMBEED, INC
Warning

A590369-1000G A590369-100G A590369-10G A590369-25G A590369-500G
76726-816EA 410 USD InStock
76726-816 76726-818 76726-820 76726-822 76726-824
Furfuryl methyl disulfide 98%
Furfuryl methyl disulphide
Precaución: for Research Use Only


Formula: C₆H₈OS₂
MDL Number: MFCD00012326
CAS Number: 57500-00-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty