Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

p-Anisidinium chloride 99%

Supplier: AMBEED, INC
Warning

A594438-100G A594438-1G A594438-25G A594438-5G
76726-870EA 227 USD InStock
76726-870 76726-872 76726-874 76726-876
p-Anisidinium chloride 99%
p-Anisidinium chloride
Precaución: for Research Use Only


Formula: C₇H₁₀ClNO
MDL Number: MFCD00036388
CAS Number: 20265-97-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty