Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Disodium L(+)-tartrate 98%

Supplier: AMBEED, INC
Warning

A595566-100G A595566-25G A595566-500G A595566-1000G
76726-888EA 43 USD InStock
76726-888 76726-890 76726-892 77669-570
Disodium L(+)-tartrate 98%
Disodium L(+)-tartrate
Precaución: for Research Use Only


Formula: C₄H₄Na₂O₆
MW: 194.05
Melting Pt: 150
Density: 1.82
MDL Number: MFCD00012468
CAS Number: 868-18-8
Merck Index: 12,08833
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty