Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(tert-Butoxycarbonyl)benzoic acid 98%

Supplier: AMBEED, INC
Warning

A602561-100MG A602561-1G A602561-250MG
76726-918EA 11 USD InStock
76726-918 76726-920 76726-922
3-(tert-Butoxycarbonyl)benzoic acid 98%
3-(tert-Butoxycarbonyl)benzoic acid
Precaución: for Research Use Only


Formula: C₁₂H₁₄O₄
MW: 222.24 g/mol
MDL Number: MFCD04037899
CAS Number: 33704-19-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty