Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-5-hexynoate 98%

Supplier: AMBEED, INC
Warning

A611576-1G A611576-250MG A611576-5G
76726-988EA 24 USD InStock
76726-988 76726-990 76726-992
Methyl-5-hexynoate 98%
Methyl-5-hexynoate
Precaución: for Research Use Only


Formula: C₇H₁₀O₂
MDL Number: MFCD00671366
CAS Number: 77758-51-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty