Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium ibandronate 98% (contains <5% H2O)

Supplier: AMBEED, INC
Danger

A612115-1G A612115-250MG A612115-5G
76726-994EA 29 USD InStock
76726-994 76726-996 76726-998
Sodium ibandronate 98% (contains <5% H2O)
Sodium ibandronate
Precaución: for Research Use Only


Formula: C₉H₂₃NO₇P₂
MDL Number: MFCD07197214
CAS Number: 138844-81-2
UN: 1759
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty