Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Boc-O-tert-butyl-L-serine dicyclohexylammonium salt 98%

Supplier: AMBEED, INC
Warning

A614160-1G A614160-25G A614160-5G
76727-010EA 17 USD InStock
76727-010 76727-012 76727-014
N-Boc-O-tert-butyl-L-serine dicyclohexylammonium salt 98%
N-Boc-O-tert-butyl-L-serine dicyclohexylammonium salt
Precaución: for Research Use Only


Formula: C₂₄H₄₆N₂O₅
MW: 442.641 g/mol
MDL Number: MFCD00065590
CAS Number: 18942-50-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty