Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-2-methylphenylboronic acid pinacol ester 98%

Supplier: AMBEED, INC
Warning

A618248-1G A618248-5G
76727-052EA 8 USD InStock
76727-052 76727-054
4-Fluoro-2-methylphenylboronic acid pinacol ester 98%
4-Fluoro-2-methylphenylboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₃H₁₈BFO₂
MW: 236.09 g/mol
MDL Number: MFCD05863911
CAS Number: 815631-56-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty